Methyl 4-chloro-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

38374-1G 38374-25G 38374-500MG 38374-5G
76942-216EA 132.62 USD
76942-216 76942-218 76942-220 76942-222
Methyl 4-chloro-3-(trifluoromethyl)benzoate ≥97%
Methyl 4-chloro-3-(trifluoromethyl)benzoate
Formula: C₉H₆ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06203691
CAS Number: 115591-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR