Ethyl 3,6-difluoro-2-hydroxybenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

38364-1G 38364-25G 38364-500MG 38364-5G
76942-200EA 379.81 USD
76942-200 76942-202 76942-204 76942-206
Ethyl 3,6-difluoro-2-hydroxybenzoate ≥97% research grade
Ethyl 3,6-difluoro-2-hydroxybenzoate
Formula: C₉H₈F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD10566810
CAS Number: 1214346-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR