Methyl 3,5-dichloro-4-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

38338-1G 38338-500MG 38338-5G
76942-188EA 587.74 USD
76942-188 76942-190 76942-192
Methyl 3,5-dichloro-4-ethoxybenzoate ≥97%
Methyl 3,5-dichloro-4-ethoxybenzoate
Formula: C₁₀H₁₀Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28128386
CAS Number: 192512-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR