tert-Butyl 3-chloro-2-methoxyisonicotinate ≥97%

Supplier: AOBChem USA

Warning

38328-1G 38328-500MG 38328-5G
76942-174EA 558.04 USD
76942-174 76942-176 76942-178
tert-Butyl 3-chloro-2-methoxyisonicotinate ≥97%
tert-Butyl 3-chloro-2-methoxyisonicotinate
Formula: C₁₁H₁₄ClNO₃
MW: 243.687 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707077
CAS Number: 2413441-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR