Methyl 4-(benzyloxy)-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38257-1G 38257-25G 38257-500MG 38257-5G
76942-162EA 296 USD
76942-162 76942-164 76942-166 76942-168
Methyl 4-(benzyloxy)-2-methylbenzoate ≥97%
Methyl 4-(benzyloxy)-2-methylbenzoate
Formula: C₁₆H₁₆O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00598376
CAS Number: 175153-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR