N-Cyclohexyl-3-fluoro-2-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

38239-25G 38239-5G
76942-144EA 453.01 USD
76942-144 76942-146
N-Cyclohexyl-3-fluoro-2-methylbenzamide ≥97%
N-Cyclohexyl-3-fluoro-2-methylbenzamide
Formula: C₁₄H₁₈FNO
Storage Temperature: Ambient
MDL Number: MFCD32632322
CAS Number: 2404734-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR