6-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

38194-1G 38194-25G 38194-5G
76942-132EA 79.57 USD
76942-132 76942-134 76942-136
6-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridin-2-amine ≥97%
6-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridin-2-amine
Formula: C₁₁H₁₃N₃
MW: 187.241 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20705020
CAS Number: 5059-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR