Methyl 2-methoxy-4,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38178-1G 38178-25G 38178-500MG 38178-5G
76942-120EA 411.63 USD
76942-120 76942-122 76942-124 76942-126
Methyl 2-methoxy-4,5-dimethylbenzoate ≥97%
Methyl 2-methoxy-4,5-dimethylbenzoate
Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17167899
CAS Number: 1245534-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR