Methyl 2-fluoro-3-propoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

37999-1G 37999-25G 37999-500MG 37999-5G
76942-044EA 126.25 USD
76942-044 76942-046 76942-048 76942-050
Methyl 2-fluoro-3-propoxybenzoate ≥97%
Methyl 2-fluoro-3-propoxybenzoate
Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632289
CAS Number: 2404733-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR