Ethyl 4-(benzyloxy)-5-bromo-2,3-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

37888-1G 37888-500MG 37888-5G
76942-016EA 653.52 USD
76942-016 76942-018 76942-020
Ethyl 4-(benzyloxy)-5-bromo-2,3-difluorobenzoate ≥97%
Ethyl 4-(benzyloxy)-5-bromo-2,3-difluorobenzoate
Formula: C₁₆H₁₃BrF₂O₃
MW: 371.173 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632272
CAS Number: 2404733-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR