7-Fluoro-1H-indazol-3-amine ≥97% research grade

Supplier: AOBChem USA

Warning

37839-1G 37839-25G 37839-5G
76942-004EA 106.09 USD
76942-004 76942-006 76942-008
7-Fluoro-1H-indazol-3-amine ≥97% research grade
7-Fluoro-1H-indazol-3-amine
Formula: C₇H₆FN₃
MW: 151.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03426694
CAS Number: 404827-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR