N-Cyclopropyl-3-methoxy-4-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

37760-1G 37760-25G 37760-500MG 37760-5G
76941-992EA 292.81 USD
76941-992 76941-994 76941-996 76941-998
N-Cyclopropyl-3-methoxy-4-methylbenzamide ≥97%
N-Cyclopropyl-3-methoxy-4-methylbenzamide
Formula: C₁₂H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD10559798
CAS Number: 1007767-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR