Methyl 5-ethylthiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

37657-1G 37657-25G 37657-500MG 37657-5G
76941-964EA 343.74 USD
76941-964 76941-966 76941-968 76941-970
Methyl 5-ethylthiophene-2-carboxylate ≥97%
Methyl 5-ethylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₀O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD06203733
CAS Number: 91086-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR