Methyl 4-fluoro-2-methoxy-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37624-1G 37624-500MG 37624-5G
76941-950EA 688.53 USD
76941-950 76941-952 76941-954
Methyl 4-fluoro-2-methoxy-3-methylbenzoate ≥97%
Methyl 4-fluoro-2-methoxy-3-methylbenzoate
Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707038
CAS Number: 2112758-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR