Ethyl 4-chloro-3,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37611-1G 37611-25G 37611-500MG 37611-5G
76941-936EA 285.39 USD
76941-936 76941-938 76941-940 76941-942
Ethyl 4-chloro-3,5-dimethylbenzoate ≥97%
Ethyl 4-chloro-3,5-dimethylbenzoate
Formula: C₁₁H₁₃ClO₂
MW: 212.673 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11554996
CAS Number: 1256466-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR