Ethyl-3-chloropicolinate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 3-chloropicolinate, Ethyl 3-chloro-2-pyridinecarboxylate

37606-1G 37606-25G 37606-500MG 37606-5G
76941-920EA 173.99 USD
76941-920 76941-922 76941-924 76941-926
Ethyl-3-chloropicolinate ≥95%
Ethyl 3-chloropicolinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD11656307
CAS Number: 128073-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR