Benzyl 2-bromo-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

37585-25G 37585-5G
76941-902EA 524.09 USD
76941-902 76941-904
Benzyl 2-bromo-5-(trifluoromethyl)benzoate ≥97%
Benzyl 2-bromo-5-(trifluoromethyl)benzoate
Formula: C₁₅H₁₀BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD32632243
CAS Number: 2404733-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR