Ethyl 2,3-dichloro-6-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

37576-1G 37576-25G 37576-5G
76941-892EA 73.21 USD
76941-892 76941-894 76941-896
Ethyl 2,3-dichloro-6-fluorobenzoate ≥97%
Ethyl 2,3-dichloro-6-fluorobenzoate
Formula: C₉H₇Cl₂FO₂
MW: 237.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698217
CAS Number: 1214332-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR