Methyl 3-(benzyloxy)-6-bromo-2-chlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

37569-1G 37569-25G 37569-500MG 37569-5G
76941-864EA 290.69 USD
76941-864 76941-866 76941-868 76941-870
Methyl 3-(benzyloxy)-6-bromo-2-chlorobenzoate ≥97%
Methyl 3-(benzyloxy)-6-bromo-2-chlorobenzoate
Formula: C₁₅H₁₂BrClO₃
MW: 355.611 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632239
CAS Number: 2404733-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR