Ethyl 6-chloro-4-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

37499-1G 37499-25G 37499-500MG 37499-5G
76941-826EA 215.37 USD
76941-826 76941-828 76941-830 76941-832
Ethyl 6-chloro-4-methylnicotinate ≥97%
Ethyl 6-chloro-4-methylnicotinate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.634 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18416512
CAS Number: 57591-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR