Ethyl 5-bromo-2,3-difluoro-4-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

37441-1G 37441-25G 37441-500MG 37441-5G
76941-810EA 409.51 USD
76941-810 76941-812 76941-814 76941-816
Ethyl 5-bromo-2,3-difluoro-4-methoxybenzoate ≥97%
Ethyl 5-bromo-2,3-difluoro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉BrF₂O₃
MW: 295.077 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632221
CAS Number: 2122218-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR