8-Methyl-[1,2,4]triazolo[4,3-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

37266-1G 37266-25G 37266-500MG 37266-5G
76941-772EA 292.81 USD
76941-772 76941-774 76941-776 76941-778
8-Methyl-[1,2,4]triazolo[4,3-a]pyridine ≥97%
8-Methyl-[1,2,4]triazolo[4,3-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.151 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09994424
CAS Number: 4997-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR