6-Bromo-2-chloro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37222-1G 37222-25G 37222-5G
76941-760EA 105.03 USD
76941-760 76941-762 76941-764
6-Bromo-2-chloro-3-methoxybenzaldehyde ≥97%
6-Bromo-2-chloro-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD32632167
CAS Number: 1358745-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR