6-Bromo-5-methyl-[1,2,4]triazolo[4,3-a]pyridine ≥95%

Supplier: AOBChem USA

Warning

37196-1G 37196-25G 37196-500MG 37196-5G
76941-744EA 192.03 USD
76941-744 76941-746 76941-748 76941-750
6-Bromo-5-methyl-[1,2,4]triazolo[4,3-a]pyridine ≥95%
6-Bromo-5-methyl-[1,2,4]triazolo[4,3-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.047 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22381236
CAS Number: 1427413-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR