Ethyl 5-bromobenzo[d][1,3]dioxole-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

37184-1G 37184-25G 37184-500MG 37184-5G
76941-722EA 350.1 USD
76941-722 76941-724 76941-726 76941-728
Ethyl 5-bromobenzo[d][1,3]dioxole-4-carboxylate ≥97%
Ethyl 5-bromobenzo[d][1,3]dioxole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₄
MW: 273.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29917949
CAS Number: 2092700-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR