Ethyl 4-(benzyloxy)-3-chlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

37181-1G 37181-25G 37181-500MG 37181-5G
76941-706EA 355.41 USD
76941-706 76941-708 76941-710 76941-712
Ethyl 4-(benzyloxy)-3-chlorobenzoate ≥97%
Ethyl 4-(benzyloxy)-3-chlorobenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₅ClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD06204509
CAS Number: 50529-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR