Ethyl 4-methoxy-3,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37176-1G 37176-25G 37176-500MG 37176-5G
76941-694EA 350.1 USD
76941-694 76941-696 76941-698 76941-700
Ethyl 4-methoxy-3,5-dimethylbenzoate ≥97%
Ethyl 4-methoxy-3,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11555029
CAS Number: 105401-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR