tert-Butyl 2-fluoro-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

36904-1G 36904-25G 36904-500MG 36904-5G
76941-634EA 221.73 USD
76941-634 76941-636 76941-638 76941-640
tert-Butyl 2-fluoro-3-(trifluoromethyl)benzoate ≥97%
tert-Butyl 2-fluoro-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₂F₄O₂
MW: 264.216 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22906597
CAS Number: 1505194-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR