8-Bromo-[1,2,4]triazolo[4,3-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

36827-1G 36827-25G 36827-5G
76941-598EA 96.55 USD
76941-598 76941-600 76941-602
8-Bromo-[1,2,4]triazolo[4,3-a]pyridine ≥97%
8-Bromo-[1,2,4]triazolo[4,3-a]pyridine
Formula: C₆H₄BrN₃
Storage Temperature: Ambient
MDL Number: MFCD00498951
CAS Number: 1126824-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR