6-Bromo[1,2,4]triazolo[4,3-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-[1,2,4]triazolo[4,3-a]pyridine

36823-25G 36823-5G
76941-586EA 225.98 USD
76941-586 76941-588
6-Bromo[1,2,4]triazolo[4,3-a]pyridine ≥97%
6-Bromo-[1,2,4]triazolo[4,3-a]pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689506
CAS Number: 108281-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR