Methyl-3-bromo-5-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

36698-25G 36698-5G
76941-566EA 410.57 USD
76941-566 76941-568
Methyl-3-bromo-5-ethoxybenzoate ≥97%
Methyl 3-bromo-5-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
Storage Temperature: Ambient
MDL Number: MFCD16295085
CAS Number: 860695-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR