Ethyl 6-methoxy-2-naphthoate ≥97%

Supplier: AOBChem USA

Warning

36673-1G 36673-25G 36673-500MG 36673-5G
76941-544EA 144.29 USD
76941-544 76941-546 76941-548 76941-550
Ethyl 6-methoxy-2-naphthoate ≥97%
Ethyl 6-methoxy-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00841138
CAS Number: 6297-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR