Ethyl 5-chloro-2,4-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

36583-1G 36583-25G 36583-500MG 36583-5G
76941-528EA 173.99 USD
76941-528 76941-530 76941-532 76941-534
Ethyl 5-chloro-2,4-difluorobenzoate ≥97%
Ethyl 5-chloro-2,4-difluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇ClF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18416425
CAS Number: 1239592-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR