Methyl-6-bromo-2-chloronicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 6-bromo-2-chloronicotinate

36414-1G 36414-25G 36414-500MG 36414-5G
76941-486EA 401.03 USD
76941-486 76941-488 76941-490 76941-492
Methyl-6-bromo-2-chloronicotinate ≥97%
Methyl-6-bromo-2-chloronicotinate
Formula: C₇H₅BrClNO₂
MW: 250.49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11857730
CAS Number: 1142192-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR