tert-Butyl 3-bromo-5-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

36357-1G 36357-25G 36357-500MG 36357-5G
76941-470EA 231.28 USD
76941-470 76941-472 76941-474 76941-476
tert-Butyl 3-bromo-5-iodobenzoate ≥97%
tert-Butyl 3-bromo-5-iodobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrIO₂
MW: 383.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22123728
CAS Number: 690260-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR