tert-Butyl 2,6-difluoronicotinate ≥97%

Supplier: AOBChem USA

Warning

36253-1G 36253-25G 36253-5G
76941-432EA 67.9 USD
76941-432 76941-434 76941-436
tert-Butyl 2,6-difluoronicotinate ≥97%
tert-Butyl 2,6-difluoronicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₂NO₂
MW: 215.197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29923050
CAS Number: 1956375-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR