Ethyl (4-bromo-1H-pyrazol-1-yl)acetate ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(4-bromo-1H-pyrazol-1-yl)acetate

36228-1G 36228-25G 36228-5G
76941-422EA 64.72 USD
76941-422 76941-424 76941-426
Ethyl (4-bromo-1H-pyrazol-1-yl)acetate ≥95% research grade
Ethyl (4-bromo-1H-pyrazol-1-yl)acetate
Formula: C₇H₉BrN₂O₂
MW: 233.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04969156
CAS Number: 82231-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR