Methyl 4-bromo-5-methylthiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

36217-1G 36217-25G 36217-500MG 36217-5G
76941-406EA 352.22 USD
76941-406 76941-408 76941-410 76941-412
Methyl 4-bromo-5-methylthiophene-2-carboxylate ≥97%
Methyl 4-bromo-5-methylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇BrO₂S
MW: 235.098 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14584477
CAS Number: 237385-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR