Methyl 2-chloro-3,5-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

36140-1G 36140-25G 36140-500MG 36140-5G
76941-390EA 266.29 USD
76941-390 76941-392 76941-394 76941-396
Methyl 2-chloro-3,5-difluorobenzoate ≥97%
Methyl 2-chloro-3,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅ClF₂O₂
MW: 206.574 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18417405
CAS Number: 1261846-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR