5-Methyl-1H-indazole-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

35897-1G 35897-500MG 35897-5G
76941-346EA 736.27 USD
76941-346 76941-348 76941-350
5-Methyl-1H-indazole-4-carboxylic acid ≥97%
5-Methyl-1H-indazole-4-carboxylic acid
Formula: C₉H₈N₂O₂
MW: 176.1720 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22556693
CAS Number: 1360927-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR