tert-Butyl 4-iodo-3-methyl-benzoate ≥97%

Supplier: AOBChem USA

Warning

35867-1G 35867-25G 35867-5G
76941-334EA 62.6 USD
76941-334 76941-336 76941-338
tert-Butyl 4-iodo-3-methyl-benzoate ≥97%
tert-Butyl 4-iodo-3-methyl-benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅IO₂
MW: 318.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631840
CAS Number: 360059-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR