N-Methyl-4-bromo-3-Methylbenzamide ≥97%
Supplier: AOBChem USA
|
Warning
|
35847-1G
35847-25G
35847-500MG
35847-5G
76941-306EA
120.95
USD
76941-306
76941-308
76941-310
76941-312
N-Methyl-4-bromo-3-Methylbenzamide ≥97%
N-Methyl-4-bromo-3-Methylbenzamide
Precaución: for Research Use Only
Formula:
C₉H₁₀BrNO Storage Temperature: Ambient |
MDL Number:
MFCD26802222 CAS Number: 149104-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...