Methyl-3-methoxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 3-methoxy-5-methylbenzoate

35689-25G 35689-5G
76941-286EA 516.66 USD
76941-286 76941-288
Methyl-3-methoxy-5-methylbenzoate ≥97%
Methyl 3-methoxy-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09833214
CAS Number: 108593-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR