Methyl 3-bromo-2-chloro-6-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

35612-1G 35612-25G 35612-500MG 35612-5G
76941-272EA 132.62 USD
76941-272 76941-274 76941-276 76941-278
Methyl 3-bromo-2-chloro-6-fluorobenzoate ≥95%
Methyl 3-bromo-2-chloro-6-fluorobenzoate
Formula: C₈H₅BrClFO₂
MW: 267.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28140551
CAS Number: 1784053-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR