2-Chloro-4-methylnicotinic acid ethyl ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-chloro-4-methylnicotinate

35561-1G 35561-25G 35561-500MG 35561-5G
76941-256EA 242.95 USD
76941-256 76941-258 76941-260 76941-262
2-Chloro-4-methylnicotinic acid ethyl ester ≥97%
2-Chloro-4-methylnicotinic acid ethyl ester

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11501333
CAS Number: 50840-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR