6-Chloro-2-fluoro-3-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

35393-1G 35393-25G 35393-5G
76941-186EA 62.6 USD
76941-186 76941-188 76941-190
6-Chloro-2-fluoro-3-methoxyphenylboronic acid pinacol ester ≥97%
6-Chloro-2-fluoro-3-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BClFO₃
MW: 286.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31390766
CAS Number: 1628684-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR