Methyl 6-bromo-5-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

35337-1G 35337-25G 35337-500MG 35337-5G
76941-170EA 260.99 USD
76941-170 76941-172 76941-174 76941-176
Methyl 6-bromo-5-methylnicotinate ≥97%
Methyl 6-bromo-5-methylnicotinate
Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18258594
CAS Number: 1210451-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR