6-Chloro-8-fluoroquinoline ≥97%

Supplier: AOBChem USA

Warning

35297-1G 35297-25G 35297-5G
76941-162EA 88.06 USD
76941-162 76941-164 76941-166
6-Chloro-8-fluoroquinoline ≥97%
6-Chloro-8-fluoroquinoline
Formula: C₉H₅ClFN
Storage Temperature: Refrigerator
MDL Number: MFCD16658595
CAS Number: 52200-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR