Ethyl 2-fluoro-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

35070-1G 35070-25G 35070-500MG 35070-5G
76941-118EA 215.37 USD
76941-118 76941-120 76941-122 76941-124
Ethyl 2-fluoro-5-methoxybenzoate ≥97%
Ethyl 2-fluoro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204417
CAS Number: 773134-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR