Ethyl 4-bromo-2,6-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

35038-1G 35038-25G 35038-500MG 35038-5G
76941-092EA 135.8 USD
76941-092 76941-094 76941-096 76941-098
Ethyl 4-bromo-2,6-dimethylbenzoate ≥97%
Ethyl 4-bromo-2,6-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD28738476
CAS Number: 1807169-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR